Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mab-20
Entrez Gene ID
171727
Gene Name
protein-coding
Gene Aliases
CELE_Y71G12B.20
Chromosome Location
Chr.I: 1644588 - 1651457 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058474.5 NP_490875.1 1 - 2 279 99
NR_165326.1 1 - 2 77 99

View Full Product Details