Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mab-20
Entrez Gene ID
171727
Gene Name
protein-coding
Gene Aliases
CELE_Y71G12B.20
Chromosome Location
Chr.I: 1644588 - 1651457 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306514.3 NP_001293443.1 3 - 4 628 76
NM_001306515.3 NP_001293444.1 2 - 3 457 76
NM_058474.5 NP_490875.1 4 - 5 1190 76
NR_165326.1 3 - 4 1875 76

View Full Product Details