Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lpd-6
Entrez Gene ID
171886
Gene Name
Brix domain-containing protein
Gene Aliases
CELE_K09H9.6
Chromosome Location
Chr.I: 3144410 - 3147793 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058707.5 NP_491108.2 3 - 4 687 131

View Full Product Details