Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
xnp-1
Entrez Gene ID
172077
Gene Name
Transcriptional regulator ATRX homolog
Gene Aliases
CELE_B0041.7
Chromosome Location
Chr.I: 4665666 - 4671738 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025787.4 NP_001020958.1 7 - 8 2927 161

View Full Product Details