Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
frm-4
Entrez Gene ID
172189
Gene Name
protein-coding
Gene Aliases
CELE_C24A11.8
Chromosome Location
Chr.I: 5402882 - 5408344 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001262958.3 NP_001249887.1 4 - 5 304 102
NM_001383090.1 4 - 5 306 102

View Full Product Details