Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
npp-7
Entrez Gene ID
172234
Gene Name
Nuclear Pore complex Protein
Gene Aliases
CELE_T19B4.2
Chromosome Location
Chr.I: 5692264 - 5697506 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383135.1 3 - 4 967 105

View Full Product Details