Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tyr-4
Entrez Gene ID
172263
Gene Name
TYRosinase
Gene Aliases
CELE_C34G6.2
Chromosome Location
Chr.I: 5898303 - 5902118 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059308.8 NP_491709.1 6 - 7 1184 66

View Full Product Details