Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mlk-1
Entrez Gene ID
178895
Gene Name
protein-coding
Gene Aliases
CELE_K11D12.10
Chromosome Location
Chr.V: 5056367 - 5064811 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313222.3 NP_001300151.1 1 - 2 77 94
NM_001313223.3 NP_001300152.1 1 - 1 49 94
NM_171456.8 NP_741536.1 1 - 2 83 94
NM_171928.6 NP_741537.1 1 - 2 75 94

View Full Product Details