Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-63
Entrez Gene ID
180068
Gene Name
Nuclear Hormone Receptor family
Gene Aliases
CELE_C06C6.4
Chromosome Location
Chr.V: 16002588 - 16004735 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171557.5 NP_741645.3 5 - 6 739 107

View Full Product Details