Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gbb-1
Entrez Gene ID
189837
Gene Name
protein-coding
Gene Aliases
CELE_Y41G9A.4
Chromosome Location
Chr.X: 2957934 - 2968194 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270068.6 NP_001256997.2 16 - 17 2320 77
NM_001313456.4 NP_001300385.1 17 - 17 2335 77
NM_001313457.4 NP_001300386.1 2 - 3 394 77
NM_171645.6 NP_741740.3 17 - 18 2451 77
GenBank mRNA EU729334.1 - 16 - 17 2302 77

View Full Product Details