Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pccb-1
Entrez Gene ID
180596
Gene Name
protein-coding
Gene Aliases
CELE_F52E4.1
Chromosome Location
Chr.X: 3089496 - 3092457 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171648.4 NP_741743.1 1 - 2 67 64

View Full Product Details