Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gly-12
Entrez Gene ID
181005
Gene Name
GLYcosylation related
Gene Aliases
CELE_F48E3.1
Chromosome Location
Chr.X: 7485864 - 7490707 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171727.4 NP_741838.1 9 - 10 1344 68
GenBank mRNA AF082011.1 - 9 - 10 1344 68

View Full Product Details