Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-14
Entrez Gene ID
181102
Gene Name
protein-coding
Gene Aliases
CELE_T01B10.4
Chromosome Location
Chr.X: 8473722 - 8476178 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171742.5 NP_741860.2 5 - 6 1256 69

View Full Product Details