Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ikke-1
- Entrez Gene ID
-
176278
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_R107.4
- Chromosome Location
-
Chr.III: 9046039 - 9050700 on Build WBcel235
- Gene Symbol
- R107.2
- Entrez Gene ID
-
176277
- Gene Name
- ATP-dependent (S)-NAD(P)H-hydrate dehydratase
- Gene Aliases
-
CELE_R107.2
- Chromosome Location
-
Chr.III: 9043886 - 9050693 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_066600.5 |
NP_499001.1 |
2 - 3 |
602 |
114 |
View Full Product Details