Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
clp-4
Entrez Gene ID
175326
Gene Name
protein-coding
Gene Aliases
CELE_Y39A3CL.5
Chromosome Location
Chr.III: 1807730 - 1818325 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065059.9 NP_497460.2 1 - 2 57 100
NM_181914.6 NP_871643.1 1 - 2 59 100

View Full Product Details