Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gly-15
Entrez Gene ID
173114
Gene Name
protein-coding
Gene Aliases
CELE_C54C8.11
Chromosome Location
Chr.I: 12462176 - 12465816 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060763.4 NP_493164.2 3 - 4 484 123

View Full Product Details