Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
kal-1
Entrez Gene ID
173282
Gene Name
human KALlmann syndrome homolog
Gene Aliases
CELE_K03D10.1
Chromosome Location
Chr.I: 14128587 - 14142207 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061067.8 NP_493468.2 1 - 2 106 75

View Full Product Details