Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
npp-4
Entrez Gene ID
173329
Gene Name
Nuclear Pore complex Protein
Gene Aliases
CELE_Y54E5A.4
Chromosome Location
Chr.I: 14701118 - 14706522 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061151.5 NP_493552.1 4 - 5 1396 67

View Full Product Details