Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sur-2
Entrez Gene ID
173345
Gene Name
protein-coding
Gene Aliases
CELE_F39B2.4
Chromosome Location
Chr.I: 14777137 - 14787804 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026292.5 NP_001021463.1 14 - 15 3762 93
NM_061174.6 NP_493575.2 14 - 15 3757 93
GenBank mRNA U33051.1 - 14 - 15 3758 93

View Full Product Details