Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srt-54
Entrez Gene ID
187250
Gene Name
Serpentine Receptor, class T
Gene Aliases
CELE_K10B4.2
Chromosome Location
Chr.II: 117216 - 118691 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061264.1 NP_493665.1 3 - 4 726 85

View Full Product Details