Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clec-43
Entrez Gene ID
187664
Gene Name
C-type LECtin
Gene Aliases
CELE_R07C3.1
Chromosome Location
Chr.II: 944066 - 946157 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061458.3 NP_493859.2 1 - 2 151 91

View Full Product Details