Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
col-40
Entrez Gene ID
173495
Gene Name
Cuticle collagen 40
Gene Aliases
CELE_T13B5.4
Chromosome Location
Chr.II: 1104942 - 1106967 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061512.4 NP_493913.2 3 - 4 252 66

View Full Product Details