Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- math-45
- Entrez Gene ID
-
173526
- Gene Name
- MATH (meprin-associated Traf homology) domain containing
- Gene Aliases
-
CELE_Y51H7BM.1
- Chromosome Location
-
Chr.II: 1510287 - 1517454 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_061582.6 |
NP_493983.4 |
3 - 4 |
588 |
137 |
View Full Product Details