Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
math-45
Entrez Gene ID
173526
Gene Name
MATH (meprin-associated Traf homology) domain containing
Gene Aliases
CELE_Y51H7BM.1
Chromosome Location
Chr.II: 1510287 - 1517454 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061582.6 NP_493983.4 7 - 8 1396 65

View Full Product Details