Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxc-49
Entrez Gene ID
188543
Gene Name
F-box C protein
Gene Aliases
CELE_T16A1.3
Chromosome Location
Chr.II: 2084129 - 2085213 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061775.2 NP_494176.1 2 - 3 368 149

View Full Product Details