Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bath-9
Entrez Gene ID
173681
Gene Name
BTB and MATH domain containing
Gene Aliases
CELE_Y49F6C.3
Chromosome Location
Chr.II: 3377089 - 3378834 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062123.4 NP_494524.1 2 - 3 515 101

View Full Product Details