Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srg-28
Entrez Gene ID
353464
Gene Name
Serpentine receptor class gamma
Gene Aliases
CELE_T09D3.2
Chromosome Location
Chr.V: 5334888 - 5336329 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182427.1 NP_872227.1 1 - 2 229 77

View Full Product Details