Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rig-6
Entrez Gene ID
173828
Gene Name
protein-coding
Gene Aliases
CELE_C33F10.5
Chromosome Location
Chr.II: 4764147 - 4812086 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026843.7 NP_001022014.1 4 - 5 809 73

View Full Product Details