Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mpk-2
Entrez Gene ID
173880
Gene Name
protein-coding
Gene Aliases
CELE_C04G6.1
Chromosome Location
Chr.II: 5104118 - 5108901 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026742.5 NP_001021913.1 9 - 10 1395 102
NM_062545.8 NP_494946.2 9 - 10 1374 102

View Full Product Details