Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-22
Entrez Gene ID
174066
Gene Name
Nuclear hormone receptor family member nhr-22
Gene Aliases
CELE_K06A1.4
Chromosome Location
Chr.II: 6446195 - 6449815 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062897.6 NP_495298.1 7 - 8 1509 106

View Full Product Details