Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
aff-1
Entrez Gene ID
174123
Gene Name
Cell fusion protein aff-1
Gene Aliases
CELE_C44B7.3
Chromosome Location
Chr.II: 6860582 - 6863752 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063001.6 NP_495402.3 6 - 7 910 99
GenBank mRNA EF205023.1 - 6 - 7 910 99

View Full Product Details