Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gld-3
Entrez Gene ID
174174
Gene Name
protein-coding
Gene Aliases
CELE_T07F8.3
Chromosome Location
Chr.II: 7132643 - 7137522 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063078.4 NP_495479.2 8 - 9 885 111
NM_063079.7 NP_495480.1 8 - 9 885 111

View Full Product Details