Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sro-1
Entrez Gene ID
191958
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_D1022.6
Chromosome Location
Chr.II: 7460026 - 7465619 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001377819.1 NP_001364737.1 2 - 3 156 100

View Full Product Details