Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-105
Entrez Gene ID
174306
Gene Name
protein-coding
Gene Aliases
CELE_C41C4.5
Chromosome Location
Chr.II: 8116459 - 8122914 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129123.3 NP_001122595.1 4 - 5 754 67
NM_001267210.3 NP_001254139.1 5 - 6 715 67
NM_001377828.2 NP_001364812.1 5 - 6 742 67
NM_001381517.1 NP_001368112.1 5 - 6 736 67
NM_001381518.2 NP_001367274.1 5 - 6 736 67
NM_063301.6 NP_495702.1 4 - 5 554 67

View Full Product Details