Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rpl-10
Entrez Gene ID
174311
Gene Name
60S ribosomal protein L10
Gene Aliases
CELE_F10B5.1
Chromosome Location
Chr.II: 8143881 - 8144683 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063306.8 NP_495707.1 1 - 2 243 62

View Full Product Details