Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
qua-1
Entrez Gene ID
174319
Gene Name
Protein qua-1
Gene Aliases
CELE_T05C12.10
Chromosome Location
Chr.II: 8198803 - 8204005 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063324.4 NP_495725.2 1 - 2 89 102

View Full Product Details