Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lact-4
Entrez Gene ID
174356
Gene Name
Beta-lactamase domain-containing protein
Gene Aliases
CELE_M05D6.4
Chromosome Location
Chr.II: 8473839 - 8482218 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063389.7 NP_495790.1 6 - 7 1430 77

View Full Product Details