Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
suf-1
Entrez Gene ID
174380
Gene Name
Suf domain-containing protein
Gene Aliases
CELE_F28C6.6
Chromosome Location
Chr.II: 8603858 - 8606744 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063424.7 NP_495825.1 1 - 2 91 107

View Full Product Details