Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
let-23
Entrez Gene ID
174462
Gene Name
protein-coding
Gene Aliases
CELE_ZK1067.1
Chromosome Location
Chr.II: 9197512 - 9207213 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136358.4 NP_001129830.1 10 - 11 1889 112
NM_001136359.5 NP_001129831.1 12 - 13 2087 112
NM_001313678.3 NP_001300607.1 13 - 14 2114 112
NM_063561.5 NP_495962.2 12 - 13 2161 112

View Full Product Details