Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nlt-1
Entrez Gene ID
191734
Gene Name
Non-specific lipid-transfer protein-like 1
Gene Aliases
CELE_ZK892.2
Chromosome Location
Chr.II: 10000335 - 10001279 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063760.5 NP_496161.1 2 - 3 188 81

View Full Product Details