Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clec-60
Entrez Gene ID
191384
Gene Name
VWFA domain-containing protein
Gene Aliases
CELE_ZK666.6
Chromosome Location
Chr.II: 10480043 - 10483138 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063858.3 NP_496259.1 5 - 6 644 99

View Full Product Details