Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mab-10
Entrez Gene ID
187916
Gene Name
Uncharacterized protein
Gene Aliases
CELE_R166.1
Chromosome Location
Chr.II: 10528084 - 10534380 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063867.3 NP_496268.2 8 - 9 1930 66
NR_138506.1 7 - 8 1671 66

View Full Product Details