Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- lact-3
- Entrez Gene ID
-
174644
- Gene Name
- Beta-lactamase domain-containing protein
- Gene Aliases
-
CELE_M28.6
- Chromosome Location
-
Chr.II: 10652353 - 10657841 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_063898.4 |
NP_496299.1 |
1 - 2 |
328 |
84 |
- Gene Symbol
- M28.5
- Entrez Gene ID
-
174645
- Gene Name
- NHP2-like protein 1 homolog
- Gene Aliases
-
CELE_M28.5
- Chromosome Location
-
Chr.II: 10648892 - 10658855 on Build WBcel235
View Full Product Details