Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
glb-4
Entrez Gene ID
182480
Gene Name
GLoBin related
Gene Aliases
CELE_C09H10.8
Chromosome Location
Chr.II: 11109571 - 11111207 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063981.4 NP_496382.2 3 - 4 462 72

View Full Product Details