Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
srh-44
Entrez Gene ID
181928
Gene Name
Serpentine Receptor, class H
Gene Aliases
CELE_B0334.7
Chromosome Location
Chr.II: 11515075 - 11518154 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064059.2 NP_496460.2 1 - 2 180 61

View Full Product Details