Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
patr-1
Entrez Gene ID
174808
Gene Name
Protein PAT1 homolog 1
Gene Aliases
CELE_F43G6.9
Chromosome Location
Chr.II: 11774664 - 11777943 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064113.6 NP_496514.1 4 - 5 581 103

View Full Product Details