Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nhr-61
Entrez Gene ID
175142
Gene Name
protein-coding
Gene Aliases
CELE_W01D2.2
Chromosome Location
Chr.II: 14816136 - 14840009 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027210.7 NP_001022381.1 3 - 4 384 67
NM_001027211.6 NP_001022382.1 2 - 3 246 67

View Full Product Details