Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tag-51
Entrez Gene ID
175245
Gene Name
Uncharacterized protein
Gene Aliases
CELE_K02F3.1
Chromosome Location
Chr.III: 854221 - 860072 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064876.9 NP_497277.2 3 - 4 1093 72

View Full Product Details