Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxa-67
Entrez Gene ID
175258
Gene Name
F-box domain-containing protein
Gene Aliases
CELE_T12B5.11
Chromosome Location
Chr.III: 948493 - 950670 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064901.6 NP_497302.1 2 - 3 310 108

View Full Product Details