Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
uaf-1
Entrez Gene ID
175270
Gene Name
protein-coding
Gene Aliases
CELE_Y92C3B.2
Chromosome Location
Chr.III: 1062794 - 1076555 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027796.4 NP_001022967.1 6 - 7 1362 57
NM_001381724.1 NP_001367384.1 1 - 2 303 57
GenBank mRNA U79157.1 - 6 - 7 1362 57

View Full Product Details